Would you like to log in again?
Hydrazine CHEMets Refill, CHEMetrics
Supplier: CHEMetrics
Titrets®
R-5005
CXR-5005PK 55.31 USD InStock
CXR-5005
Hydrazine CHEMets Refill, CHEMetrics
Environmental Test Kits
Test kits employ the PDMAB, paradimethylaminobenzaldehyde chemistry. PDMAB in acid solution reacts with hydrazine to form a yellow product.
Results are expressed as ppb (ug/L) or ppm (mg/L) N₂H₄.
Hydrazine is a powerful reducing agent that is used in various chemical processes and in boiler water as an oxygen scavenger. To control corrosion, residual hydrazine typically is maintained in the 0.05 to 0.1 mg/L range. Higher levels may be used to guard against corrosion when the boiler is out of service for an extended period.
Delivery information: This product contains 30 self-filling CHEMets ampoules to refill kit K-5005 for the analysis of hydrazine. R-5005 is compatible with C-5005 comparators.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty