Would you like to log in again?
VWR® Variable-Speed Peristaltic Pumps
Supplier: Avantor
VWR®
3384VW 3385VW 3389VW 3386VW
MFLX90010-88EA 331 USD InStock
MFLX90010-88 MFLX90027-63 MFLX90027-64 MFLX90027-65
VWR® Variable-Speed Peristaltic Pumps
Pumps Peristaltic Pumps
Compact, variable-flow, bi-directional, self-priming, peristaltic pumps offer precise flow deliveries
  • Digital display
  • Flow rates can vary from a low of 0.005 ml/min. to as much as 600 ml/min., depending on the pump model
  • Ideal for use with conductivity flow-thru cells, liquid chromatography, collecting fractions, pH/circulating fluids or buffers in baths, and moving corrosive materials
  • Four different tubing sizes ( 1/16", 3/32", 3/16", 1/4") and corresponding fittings/nipples provide fine resolution with a wide flow range; revolution of one roller delivers a precisely measured volume specific to the tubing size and motor speed
  • Fluid contacts only tubing for contamination-free pumping
  • Tubing may be used with fluid temperatures from –80 to 500 °F (–62 to 260 °C)
  • May be used with up to 120 feet of tubing for remote sampling
Use with food, pharmaceuticals, and other critical solutions. The tubing may be sterilized by autoclave. Unit pumps liquids and gases. Pumping dry does not harm the pump. Three rollers reduce flow pulsation, prevent siphoning, and eliminate the need for check valves. There are no valves to clog, no seals to leak. A universal wall power supply is included.
They provide outstanding flow control and flexibility for transferring and dosing liquids.
Ordering information: Supplied with silicone tubing, and polypropylene fittings/nipples, and a universal wall power supply. An adapter (not included) is needed to use battery power; please contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty