Would you like to log in again?
VWR® Check Valves
Supplier: Avantor
VWR®
MAKEUP MAKEUP MAKEUP MAKEUP
MFLX90028-05EA 44 USD InStock
MFLX90028-05 MFLX90028-06 MFLX90028-07 MFLX90028-04
VWR® Check Valves
Valves Vacuum Valves
Barbed check valves prevent damage to equipment, pumps, and flow systems by preventing backflow.
  • Chemical-resistant resin body and valve
  • Inert, high-purity fitting
  • Autoclavable
Chemical-resistant Kynar® resin body and Viton® resin valve ensure an inert, high-purity fitting that provides a smooth and clear flow path. Can be used with liquids and gases in chemical processing, pressure, and vacuum applications. Suitable for use with acids, alcohols, bases, and water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty