Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rifampicin 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Rifamycin AMP3-(4-Methylpiperazinyliminomethyl)rifamycin SVRifampin

45562-0010 45562-0050 45562-0250
TS45562-0050EA 110.14 USD InStock
TS45562-0050 TS45562-0010 TS45562-0250
Rifampicin 95%
Rifampicin


Formula: C₄₃H₅₈N₄O₁₂
MW: 822.95 g/mol
Melting Pt: 183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151389
CAS Number: 13292-46-1
Order Now
Specification Test Results
Appearance Light orange to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (100 mg, 60°C, 4 h) (vacuum)
HPLC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty