Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Homoserine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-DL-HomoSer-OH(±)-HomoserineH-DL-HoSer-OHH-DL-Hser-OH

20476-0050
TS20476-0050EA 91 USD InStock
TS20476-0050
DL-Homoserine 99%
DL-Homoserine


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 188…189 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002618
CAS Number: 1927-25-9
EINECS: 217-661-8
Order Now
Specification Test Results
Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Solubility (1 % in water) Clear colorless
Titration with HClO4 98.5 to 101.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty