Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hydroxyindol-3-ylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(5-Hydroxy-1H-indol-3-yl)acetic acid5-Hydroxyindole-3-acetic acid

12128-0010 12128-2500 12128-0050
TS12128-2500EA 51 USD InStock
TS12128-2500 TS12128-0010 TS12128-0050
5-Hydroxyindol-3-ylacetic acid 99%
5-Hydroxyindol-3-ylacetic acid


Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005639
CAS Number: 54-16-0
EINECS: 200-195-4
Order Now
Specification Test Results
Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 162 to 165 °C
Water ≤0.5 % (K.F.)
Titration with NaOH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty