Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Alanine 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: D(-)-Alanineα-Alanine(R)-(-)-AlanineAlanineD-α-AlanineH-D-Ala-OH

17208-0250 17208-1000 17208-0050
TS17208-0050EA 49 USD InStock
TS17208-0050 TS17208-0250 TS17208-1000
D-Alanine 99+%
D-Alanine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 278…282 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 hrs)
Sulfated ash ≤0.2 %
HPLC ≤1 % (L-content)
UV transmittance ≥95 % (c=10, H2O, 430 nm) 1 cm cell
Specific optical rotation -13.5° to -15.5° (20°C, 589 nm) (c=10, 6 N HCl) (on dry substance)
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤0.1 %
Arsenic (As) ≤2 ppm (As2O3)
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty