Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Alanine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: α-Alanine(±)-AlanineH-DL-Ala-OHDL-α-Alanine

15909-5000 15909-1000
TS15909-1000EA 31 USD InStock
TS15909-1000 TS15909-5000
DL-Alanine 99%
DL-Alanine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 272…275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (C=10, H2O, 430 nm) 1 cm cell
Titration with HClO4 98.5 to 101.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤20 ppm
Arsenic oxide (As2O3) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty