Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium alginate

Supplier: Thermo Scientific Chemicals
Synonyms: Alginic acid sodium salt

17777-2500 17777-5000 17777-0025 17777-0050
TS17777-5000EA 164.94 USD InStock
TS17777-5000 TS17777-0050 TS17777-2500 TS17777-0025
Sodium alginate
Sodium alginate


Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Order Now
Specification Test Results
Appearance Off-white to yellow-brown Powder
Infrared spectrum Conforms
Loss on drying ≤15.0 % (100 mg, 105°C, 4 hrs)
Sulfated ash 30.0 % to 36.0 % (on dried substance)
Viscosity 350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
pH 5.5 to 8.0 (1 % at 20°C)
Heavy metals (as Pb) ≤20 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty