Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Galactitol 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: Dulcitol

11770-5000 11770-0250 11770-1000
TS11770-0250EA 55 USD InStock
TS11770-0250 TS11770-1000 TS11770-5000
Galactitol 99+%
Galactitol


Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 275…280 °C (1 mmHg)
Melting Pt: 188…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00064288
CAS Number: 608-66-2
EINECS: 210-165-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 187 to 191 °C
Water ≤1.0 % (K.F.)
Solubility (3 % in water) Colorless
HPLC ≥99.0 % (ELSD)
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.01 % (in dilute NH4OH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty