Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',3'-Dideoxycytidine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

44950-1000 44950-5000
TS44950-5000EA 392 USD InStock
TS44950-5000 TS44950-1000
2',3'-Dideoxycytidine 98%
2',3'-Dideoxycytidine


Formula: C₉H₁₃N₃O₃
MW: 211.22 g/mol
Melting Pt: 216…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012188
CAS Number: 7481-89-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty