Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vincristine sulfate 95%

Supplier: Thermo Scientific Chemicals
Danger

20344-0050
TS20344-0050EA 194 USD InStock
TS20344-0050
Vincristine sulfate 95%
Vincristine sulphate


Formula: C₄₆H₅₈N₄O₁₄S
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00084729
CAS Number: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤12 %
HPLC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty