Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: H-D-Ser-OMe.HClD-Serine methyl ester hydrochlorideH-D-Ser-OMe·HCl

39679-0250
TS39679-0250EA 136 USD InStock
TS39679-0250
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 98%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride


Formula: C₄H₉O₃N·HCl
MW: 155.6 g/mol
Melting Pt: 152…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066121
CAS Number: 5874-57-7
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Specific optical rotation -2.5° to -4.5° (20°C, 589 nm) (C=4, methanol)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty