Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-HomoArg-OH.HClL(+)-Homoarginine hydrochlorideH-HomoArg-OH·HCl

16909-0050 16909-0010
TS16909-0050EA 230 USD InStock
TS16909-0050 TS16909-0010
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 98%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride


Formula: C₇H₁₆N₄O₂·HCl
MW: 224.7 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012622
CAS Number: 1483-01-8
Order Now
Specification Test Results
Appearance White Granular powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Specific optical rotation +17° to +19° (20°C, 589 nm) (c=1, 1 N HCl)
Titration with HClO4 ≥97.5 % (On dried substance)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty