Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Leu-OMe.HClL-Leucine methyl ester hydrochlorideH-Leu-OMe·HCl

12513-0250
TS12513-0250EA 70.36 USD InStock
TS12513-0250
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride


Formula: C₇H₁₅NO₂·HCl
MW: 181.7 g/mol
Melting Pt: 145…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012494
CAS Number: 7517-19-3
Order Now
Specification Test Results
Appearance White to yellowish-white Powder or crystals
Infrared spectrum Conforms
Specific optical rotation +18.5° to +21.5° (20°C, 589 nm) (c=4.5, methanol)
Titration with HClO4 ≥97.5 %
Residue after evaporation ≤0.005 %
UV at 200 nm A: ≤0.75
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty