Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clarithromycin 97%

Supplier: Thermo Scientific Chemicals
Warning

45522-0010
TS45522-0010EA 328 USD InStock
TS45522-0010
Clarithromycin 97%
Clarithromycin


Formula: C₃₈H₆₉NO₁₃
Melting Pt: 222…231 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865140
CAS Number: 81103-11-9
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 222 to 231 °C
Water ≤2 %
HPLC ≥96.0 %
Specific optical rotation -87° to -97° (20 °C, 589 nm) (c=1,CHCl3)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty