Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clotrimazole

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-[(2-Chlorophenyl)diphenylmethyl]imidazole1-(2-Chlorotrityl)imidazole

45546-1000 45546-0250 45546-0050
TS45546-1000EA 746 USD InStock
TS45546-1000 TS45546-0250 TS45546-0050
Clotrimazole
Clotrimazole


Formula: C₂₂H₁₇ClN₂
MW: 344.84 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00057220
CAS Number: 23593-75-1
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 142 to 152 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty