Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Arginine hydrochloride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Synonyms: DL-(+/-)-Arginine hydrochlorideH-DL-Arg-OH.HCl

44602-0050
TS44602-0050EA 77 USD InStock
TS44602-0050
DL-Arginine hydrochloride, anhydrous 98%
DL-Arginine hydrochloride


Formula: C₆H₁₄N₄O₂.HCl
MW: 210.66 g/mol
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00064549
CAS Number: 32042-43-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Loss on drying ≤3 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty