Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-α-Benzoyl-L-Arginine p-Nitroanilide HydrochlorideBz-Arg-pNA·HCl(S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

42864-0010
TS42864-0010EA 710 USD InStock
TS42864-0010
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride


Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Melting Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00063682
CAS Number: 21653-40-7
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +42° to +48° (20°C, 589 nm) (c=1 in DMF)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty