Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-N-Boc-amino-4-carboxytetrahydropyran 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid

36046-5000 36046-0020
TS36046-0020EA 509 USD InStock
TS36046-0020 TS36046-5000
4-N-Boc-amino-4-carboxytetrahydropyran 95%
4-N-Boc-amino-4-carboxytetrahydropyran


Formula: C₁₁H₁₉NO₅
MW: 245.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683136
CAS Number: 172843-97-9
Order Now
Specification Test Results
Appearance Off-white Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty