Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-N-3-Cyanophenylalanine 95%, 98% ee

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-Phe(3-CN)-OH

36206-0010 36206-0050
TS36206-0010EA 194 USD InStock
TS36206-0010 TS36206-0050
(S)-N-3-Cyanophenylalanine 95%, 98% ee
(S)-N-3-Cyanophenylalanine


Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860885
CAS Number: 57213-48-6
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty