Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytosine-β-D-arabinofuranoside hydrochloride 99%, crystalline

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Arabinosylcytosine hydrochlorideCytarabine HCl4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride1-(β-D-Arabinofuranosyl)cytosine HCl

22718-0010
TS22718-0010EA 276 USD InStock
TS22718-0010
Cytosine-β-D-arabinofuranoside hydrochloride 99%, crystalline
Cytosine-beta-D-arabinofuranoside hydrochloride


Formula: C₉H₁₄ClN₃O₅
MW: 279.677 g/mol
Melting Pt: 197…198 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012839
CAS Number: 69-74-9
Order Now
Specification Test Results
Appearance White to tan Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +127° to +133° (20°C, 589 nm) (c=0.7, H2O)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty