Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amikacin disulphate

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)(S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulfate)

45519-0050
TS45519-0050EA 104 USD InStock
TS45519-0050
Amikacin disulphate
Amikacin disulphate


Formula: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781.77 g/mol
Melting Pt: 220…230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167475
CAS Number: 39831-55-5
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤13 % (110°C, 3 h, 5 mm Hg)
pH 2 to 4 (1 % in water)
Titration with NaOH ≥87 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty