Would you like to log in again?
Surface Tension of Water
Supplier: ALDON CORP SE
IS7012
470338-536EA 21.25 USD InStock
470338-536
Surface Tension of Water
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Explain the principle of high surface tension as water acts like a membrane stretched across the surface preventing sulfur particles from sinking.
  • Understand and demonstrate surface tension
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A
  • Learn engineering practices: planning and carrying out investigations
A wetting agent lowers the surface tension and allows the particles to fall through the surface to the bottom of the beaker. Each kit comes with enough material to complete the experiment five times.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty