Would you like to log in again?
Autocatalysis
Supplier: ALDON CORP SE
IS7002
470338-554EA 28.5 USD InStock
470338-554
Autocatalysis
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Perform an autocatalytic reaction, observe how the product of a reaction can catalyze further reactions.
  • Understand and demonstrate autocatalysis through an autocatalytic reaction
  • Aligned to NGSS practices
Catalysis is a basic principle of chemistry and biochemistry. Some reactions are autocatylic; the product of the reaction actually catalyzes further reactions. The reaction is initiated with acid and produces acid to continue the reaction.
Caution: WARNING: This product can expose you to chemicals including strong inorganic acid mists containing sulfuric acid, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty