Would you like to log in again?
Synthetic Rubber
Supplier: ALDON CORP SE
IS7008
470338-558EA 36.9 USD InStock
470338-558
Synthetic Rubber
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Perform a synthesis reaction to produce rubber - a simple condensation polymer.
  • Understand and demonstrate polymer chemistry through a synthesis reaction
  • Aligned to NGSS practices
Show how the first synthetic rubber was made in United States by a reaction of sodium polysulfide with ethylene chloride. This produces a simple condensation polymer consisting of repeating units of ethane and polysulfide.
Caution: WARNING: This product can expose you to chemicals including ethylene dichloride (1,2-dichloroethane), which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty