Would you like to log in again?
Solid Fuel
Supplier: ALDON CORP SE
IS7025
470338-560EA 25.75 USD InStock
470338-560
Solid Fuel
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Two solutions are poured together in a beaker and form a gel. The gel can be ignited with a match. Commercially this gel is sold as Sterno®.
  • Understand and demonstrate a solid fuel synthesis reaction
  • Aligned to NGSS practices
  • Disciplinary core ideas: PS1.A, PS1.B
Caution: WARNING: This product can expose you to chemicals including methanol, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty