Would you like to log in again?
The Silicate Garden
Supplier: ALDON CORP SE
IS7031
470338-570EA 44.65 USD InStock
470338-570
The Silicate Garden
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Demonstrate chemical structures through the formation of silicate crystals.
  • Understand and demonstrate chemical crystals
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
Demonstrate polymer chemistry. Newsprint is dissolved in a solution of ammonium hydroxide and cupric sulfate. The solution is then added to a sulfuric acid solution and dark strands are formed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty