Would you like to log in again?
Oxidation of Glycerin
Supplier: ALDON CORP SE
IS7028
470338-582EA 21.79 USD InStock
470338-582
Oxidation of Glycerin
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
A small amount of potassium permanganate is placed in a petri dish. A few drops of glycerin are added and after a few seconds, a puff of smoke and violet flames are produced.
  • Understand and demonstrate an oxidation reaction
  • Aligned to NGSS practices
Perform an oxidation reaction with glycerin and potassium permanganate and witness the violet flame it produces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty