Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Q Clamp Holder

Supplier: VWR
VWR® Talon®
80862589
21572-712EA 22.85 USD InStock
21572-712
VWR® Q Clamp Holder
Clamps General Purpose Clamps
VWR® Q clamp holder for securing clamps to a rod.

  • Q clamp holder for use in custom applications
  • May be used with 21572-703 3-prong clamp head
  • Made of die cast nickel plated zinc
  • Grips support rods up to 0.5" (13 mm)
  • Threaded opening is 0.3" (7.9 mm)

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty