Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ACE20 Heavy Duty Arm Channel Mount, Grand Stands

Supplier: Grand Stands
ACE20-HD-CM-S
76538-708EA 801.61 USD InStock
76538-708
ACE20 Heavy Duty Arm Channel Mount, Grand Stands
Computer and Equipment Stands and Mounts
Single heavy duty ergonomic monitor arm with 20" height range.

  • Internal cable management
  • True sit to stand without tall ugly poles
  • 27.5" Front to back distance, compacts to 4"
  • ±45° tilt adjustment, swivels portrait to landscape
  • Up to 40 lbs. supported weight for single monitor
  • 180° (display pan swing, side to side)
  • 75×75 mm/100×100 mm

Very stable and limited bounce when the user is typing.

Ordering information: 10-year warranty.

Delivery information: Easy installation; ships assembled. Clamp and grommet mount included.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty