Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

L-Aspartyl-L-Phenylalanine

Supplier: Thermo Scientific
31002-5000 31002-1000
TS31002-5000EA 254 USD InStock
TS31002-5000 TS31002-1000
L-Aspartyl-L-Phenylalanine
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Appearance of solution: (5% in CH₃COOH 80%) clear colorless
  • Specific optical rotation: 10.0 to 13.0° (24 °C, 589 nm) (c=1, 0.5 N HCl)
  • Melting point: 236 to 239 °C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty