Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Epianhydrochlortetracycline Hydrochloride, can be used as Secondary Standard, Thermo Scientific

Supplier: Thermo Scientific
26822-1000
TS26822-1000EA 133 USD InStock
TS26822-1000
4-Epianhydrochlortetracycline Hydrochloride, can be used as Secondary Standard, Thermo Scientific
Standards Organic Standards
CAS No.: 158018-53-2

MDL No.: MFCD00143313
GHS Symbols: GHS07, GHS08
Melting Point: 222 °C (D)
Appearance: Orange Powder
Storage Temperature: Ambient
Signal word: Warning

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty