Would you like to log in again?
Thermo Scientific™ Yeast extract, for biochemistry, Thermo Scientific
Supplier: Thermo Scientific
46855-2500
TS46855-2500EA 86.84 USD InStock
TS46855-2500
Thermo Scientific™ Yeast extract, for biochemistry, Thermo Scientific
Media for Microbiology Additives and Supplements for Microbiology
  • Light yellow to yellow powder
  • Microbial contamination =<5000 CFU/g (Aerobic count)
  • Sodium chloride (NaCl) =<1.0
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty