Would you like to log in again?
ReptiFeast Live Redworms (Eisenia fetida)
Supplier: SEGREST FARMS
572-0073-3
470338-846EA 10.7 USD InStock
470338-846
ReptiFeast Live Redworms (Eisenia fetida)
Specimens Live Specimens Animals - Living Material
Red wigglers make a good live food to feed Axolotls and other aquatic amphibians and reptiles.
  • Chemical-free; safe for feeding to pets
  • Great Source of Protein
  • Also make great composting worms
ReptiFeast® live food was designed by animal keepers for animal keepers. The grab-n-go cup will make it quick and easy easy to keep alive, just store the cup refrigerated at 40°F.
Delivery information: These products contain living materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty