Would you like to log in again?
Ward's® Luria Broth with Ampicillin
Supplier: VWR International
CM279A-T9
470337-968CS 327.8 USD InStock
470337-968
Ward's® Luria Broth with Ampicillin
Media for Microbiology Dehydrated Media for Microbiology
For recombinant strains of Escherichia coli that are Ampicillin resistant.
  • Selects for ampicillin resistance
  • Sterile, no autoclaving
  • Conveniently prepared for you consistenly every time
For genetic and molecular studies using recombinant strains of Escherichia coli containing plasmids with the resistance gene to Ampicillin.
Delivery information: Prepared media will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty