Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl acetate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid allyl ester

18065-1000 18065-5000
TS18065-1000EA 48 USD InStock
TS18065-1000 TS18065-5000
Allyl acetate 99%
Allyl acetate


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 103 °C (1013 hPa)
Melting Pt: –96 °C
Density: 0.92 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008721
CAS Number: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty