Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-1-pentene 99%

Supplier: Thermo Scientific Chemicals
Danger

12740-0100 12740-0500
TS12740-0100EA 60 USD InStock
TS12740-0100 TS12740-0500
2-Methyl-1-pentene 99%
2-Methyl-1-pentene


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 62 °C (1013 hPa)
Melting Pt: –136 °C
Density: 0.68 g/cm³
Flash Pt: –26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009405
CAS Number: 763-29-1
EINECS: 212-108-7
UN: 3295
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty