Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salicylic acid 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: o-Hydroxybenzoic acid2-Hydroxybenzoic acid

41922-0100 41922-0020 41922-5000
TS41922-0100EA 1342 USD InStock
TS41922-0100 TS41922-0020 TS41922-5000
Salicylic acid 99+% ACS
Salicylic acid


Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211 °C (20 mmHg)
Melting Pt: 158…161 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 12,08484
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 158.0 to 161.0 °C
HPLC ≥99.0 %
Residue after ignition ≤0.01 %
Substance darkened by H2SO4 passes test
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty