Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butylbenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: n-Butylbenzene

10785-2500 10785-0010 10785-0500
TS10785-0010EA 486 USD InStock
TS10785-0010 TS10785-2500 TS10785-0500
Butylbenzene 99+%
Butylbenzene


Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 183 °C (760 mmHg)
Melting Pt: –88 °C
Density: 0.86 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00009463
CAS Number: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4900 (20°C, 589 nm)
GC ≥99.0 %
Methanol ≤0.5 % (GC)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty