Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Praseodymium(III,IV) oxide 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

19462-0500
TS19462-0500EA 120.29 USD InStock
TS19462-0500
Praseodymium(III,IV) oxide 99.9% (trace metals basis)
Praseodymium (III,IV) oxide


Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2012…2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9
Order Now
Specification Test Results
Appearance Dark green to dark brown to black Powder
Total conc. of imp.as REO ≤0.1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty