Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Methyl-2-butanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DL-3-Methyl-2-butanol

14993-1000 14993-5000 14993-0250
TS14993-5000EA 334 USD InStock
TS14993-5000 TS14993-1000 TS14993-0250
(±)-3-Methyl-2-butanol 98%
(±)-3-Methyl-2-butanol


Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 112 °C (760 mmHg)
Density: 0.81 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00004527
CAS Number: 598-75-4
EINECS: 209-950-2
UN: 1105
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty