Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylvaleric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Isocaproic acid4-Methylpentanoic acid

25551-0050 25551-0250
TS25551-0050EA 36 USD InStock
TS25551-0050 TS25551-0250
4-Methylvaleric acid 99%
4-Methylvaleric acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.922 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0
UN: 2922
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4136 to 1.4156 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty