Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Mercaptosuccinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Thiomalic acidDL-Mercaptosuccinic acid(±)-Mercapto-1,4-butanedioic acid

12555-0025 12555-1000 12555-5000
TS12555-1000EA 59 USD InStock
TS12555-1000 TS12555-5000 TS12555-0025
(±)-Mercaptosuccinic acid 99%
(±)-Mercaptosuccinic acid


Formula: C₄H₆O₄S
MW: 150.16 g/mol
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
Titration with NaOH 98.5 to 101.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty