Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Tolylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Methylphenyl)acetic acid2-Tolylacetic acid

13910-0250
TS13910-0250EA 63 USD InStock
TS13910-0250
o-Tolylacetic acid 99%
o-Tolylacetic acid


Formula: (C₆H₄CH₃)CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 87…90 °C
MDL Number: MFCD00004328
CAS Number: 644-36-0
EINECS: 211-416-9
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 90 °C
HPLC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty