Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N,N-diethylacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-n,n-diethylacetamideN,N,-Diethylchloroacetamide

35185-0050 35185-0250
TS35185-0250EA 88 USD InStock
TS35185-0250 TS35185-0050
2-Chloro-N,N-diethylacetamide 98%
2-Chloro-N,N-diethylacetamide


Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
Boiling Pt: 121…122 °C (12 mmHg)
Density: 1.089 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000928
CAS Number: 2315-36-8
EINECS: 219-019-2
UN: 2810
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4720 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty