Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-m-Coumaric acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2E)-3-(3-Hydroxyphenyl)acrylic acidm-Coumaric acid(E)-3-(3-Hydroxyphenyl)acrylic acidm-Hydroxycinnamic acid, predominantly transtrans-3-Hydroxycinnamic acid

12108-0250 12108-1000
TS12108-0250EA 75 USD InStock
TS12108-0250 TS12108-1000
trans-m-Coumaric acid 99%
trans-m-Coumaric acid


Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 189…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004390
CAS Number: 14755-02-3
EINECS: 209-615-0
Order Now
Specification Test Results
Appearance Beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +12° to +16° (20°C, 589 nm) (c=1, ethanol)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty