Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid 50% in water

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

45687-0250 45687-0050
TS45687-0250EA 108 USD InStock
TS45687-0250 TS45687-0050
Acetic acid 50% in water
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Flash Pt: >60 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration with NaOH 49.0 to 51.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty