Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentene 97%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propylethylenealpha-n-Amylene

43073-1000
TS43073-1000EA 139 USD InStock
TS43073-1000
1-Pentene 97%, AcroSeal™
1-Pentene


Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 30 °C (760 mmHg)
Melting Pt: –165 °C
Density: 0.64 g/cm³
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003567
CAS Number: 109-67-1
UN: 1108
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3704 to 1.3724 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty