Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

34467-0010 34467-0050
TS34467-0050EA 163 USD InStock
TS34467-0050 TS34467-0010
Methylboronic acid 97%
Methylboronic acid


Formula: CH₅BO₂
Melting Pt: 88…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002105
CAS Number: 13061-96-6
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 87 to 91 °C
Titration with NaOH 96.0 to 104.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty